Tisk…

BMS 345541 (trifluoroacetate salt) ≥98%

Dodavatel: Cayman Chemical

16667-1 16667-10 16667-25 16667-5
CAYM16667-1EA 1810 CZK
CAYM16667-1 CAYM16667-10 CAYM16667-25 CAYM16667-5
BMS 345541 (trifluoroacetate salt) ≥98%
BMS 345541 (trifluoroacetate salt)

  • A selective inhibitor of IKKα and IKKβ


A selective, cell permeable inhibitor of the IκB kinases IKKα and IKKβ (IC50s = 4 and 0.3 µM); inhibits signaling through NF-κB both in cells and in vivo, showing excellent pharmacokinetics in mice

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcda568a854b8b68e3fded8108518be26cd4bebd&vid=7c7eea0c2316178de272eb91604fdd5dd499ccc0&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR