Tisk…

Anti-CD144 Rabbit Polyclonal Antibody

Dodavatel: Cayman Chemical
160840-500
CAYM160840-500EA 34930 CZK
CAYM160840-500
Anti-CD144 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CD144
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR