Tisk…

Anti-COX-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX111]

Dodavatel: Cayman Chemical
160111-250
CAYM160111-250EA 14310 CZK
CAYM160111-250
Anti-COX-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CX111]
Protilátky
Type: Primary
Antigen: COX-1
Clonality: Monoclonal
Clone: CX111
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Human, Mouse, Sheep, Rat
Objednat

Produktová specifikace

Symbol antigenu COX-1
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Spojení FITC (Fluorescein Isothiocyanate)
Klon CX111
Reaktivita Sheep,Human,Bovine,Rat,Mouse
Hostitel Mouse
Izotyp IgG2b
ImunoFluorescence Yes
Průtoková cytometrie Yes
Imunogen ovine COX-1
Křížová adsorbce No
Skladovací pufr PBS, pH 7,2, with 50% glycerol, 0,5 mg/ml BSA and 0,02% NaN3
Skladovací teplota +4 °C
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR