Tisk…

N-Octyl-β-D-glucopyranoside

Dodavatel: Cayman Chemical
Varování

14327-1 14327-5 14327-10
CAYM14327-1EA 1810 CZK
CAYM14327-1 CAYM14327-5 CAYM14327-10
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
Vzorec: C₁₄H₂₈O₆
Bod tání: 110 °C
Storage Temperature: Freezer
MDL Number: MFCD00063288
CAS číslo: 29836-26-8
EINECS: 249-887-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR