Tisk…

Resazurin sodium salt

Dodavatel: Cayman Chemical
Varování

Synonyms: 7-Sodiooxy-3H-phenoxazin-3-one 10-oxide, Sodium 7-oxido-3-oxo-3H-phenoxazine 10-oxide, Resazurin sodium, Resazurin

14322-25 14322-10 14322-5
CAYM14322-25EA 10610 CZK
CAYM14322-25 CAYM14322-10 CAYM14322-5
Resazurin sodium salt
Resazurin sodium salt
Vzorec: C₁₂H₆NNaO₄
Storage Temperature: Ambient
MDL Number: MFCD00005036
CAS číslo: 62758-13-8
EINECS: 263-718-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR