Tisk…

Anti-DNMT Mouse Monoclonal Antibody [clone: 60B1220.1]

Dodavatel: Cayman Chemical
13479-1
CAYM13479-1EA 19580 CZK
CAYM13479-1
Anti-DNMT Mouse Monoclonal Antibody [clone: 60B1220.1]
Protilátky
Type: Primary
Antigen: DNMT
Clonality: Monoclonal
Clone: 60B1220.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Zebrafish
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR