Tisk…

Ionomycin calcium salt

Dodavatel: Cayman Chemical

11932-1 11932-5 11932-10 11932-25
CAYM11932-1EA 1350 CZK
CAYM11932-1 CAYM11932-5 CAYM11932-10 CAYM11932-25
Ionomycin calcium salt
Ionomycin calcium salt
Vzorec: C₄₁H₇₀CaO₉
MDL Number: MFCD00151183
CAS číslo: 56092-82-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR