Tisk…

5-Fluorocytosine

Dodavatel: Cayman Chemical

Synonyms: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one

11635-500 11635-1 11635-5 11635-10
CAYM11635-500EA 2000 CZK
CAYM11635-500 CAYM11635-1 CAYM11635-5 CAYM11635-10
5-Fluorocytosine
5-Fluorocytosine
Vzorec: C₄H₄FN₃O
MW: 129.09
Bod varu: 298 °C (1013 hPa)
Bod tání: 165 °C
MDL Number: MFCD00006035
CAS číslo: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR