Tisk…

7-Aminoactinomycin D (7-AAD)

Dodavatel: Cayman Chemical

11397-1 11397-5
CAYM11397-1EA 4480 CZK
CAYM11397-1 CAYM11397-5
7-Aminoactinomycin D (7-AAD)
7-Aminoactinomycin D (7-AAD)
Vzorec: C₆₂H₈₇N₁₃O₁₆
MDL Number: MFCD00077295
CAS číslo: 7240-37-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR