Tisk…

Anti-Nampt Mouse Monoclonal Antibody [clone: OMNI 379]

Dodavatel: Cayman Chemical
10813-50 10813-100
CAYM10813-50EA 11340 CZK
CAYM10813-50 CAYM10813-100
Anti-Nampt Mouse Monoclonal Antibody [clone: OMNI 379]
Protilátky
Type: Primary
Antigen: Nampt
Clonality: Monoclonal
Clone: OMNI 379
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat
Objednat

Produktová specifikace

Symbol antigenu NAMPT
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Spojení Unconjugated
Klon OMNI 379
Reaktivita Human,Rat,Mouse
Hostitel Mouse
Izotyp IgG2a kappa
ELISA Yes
Imunogen recombinant human Nampt
Křížová adsorbce No
Skladovací pufr PBS, pH 7,4
Skladovací teplota –20 °C
Transportní teplota –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR