Tisk…
HPTLC Software visionCATS
Dodavatel: CAMAG
028.0000 028.2000
CAMA028.0000EA 0 CZK
CAMA028.0000 CAMA028.2000
HPTLC Software visionCATS
Software Laboratory Software
VisionCATS is built for simplicity and intuitive operation, streamlining the entire HPTLC workflow by controlling both HPTLC PRO Modules and HPTLC instruments while efficiently managing data.
  • Comparison viewer
  • Image enhancement tools
  • Scanning densitometry and spectral evaluation
  • Quantitative analysis
  • HPTLC method library
  • Regulatory compliance
With its user-friendly interface, visionCATS guides users smoothly through the entire chromatographic process, from sample setup to report generation. Starting is easy - select a default method,
More Product Information