Tisk…

Anti-NP Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS7855
BWRLBS7855EA 10450 CZK
BWRLBS7855
Anti-NP Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human NP.

NP pAb detects endogenous levels of NP protein.

Type: Primary
Antigen: NP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Produktová specifikace

Symbol antigenu NP
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Reaktivita Human,Rat,Mouse
Hostitel Rabbit
Izotyp
Western blot Yes
Imunochemie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR