Tisk…

Anti-BAIAP2 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS7752
BWRLBS7752EA 10450 CZK
BWRLBS7752
Anti-BAIAP2 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human BAIAP2.

BAIAP2 pAb detects endogenous levels of BAIAP2 protein.

Type: Primary
Antigen: BAIAP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR