Tisk…

Anti-SIGMAR1 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS7597
BWRLBS7597EA 10450 CZK
BWRLBS7597
Anti-SIGMAR1 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human SIGMAR1.

SIGMAR1 pAb detects endogenous levels of SIGMAR1 protein.

Type: Primary
Antigen: SIGMAR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR