Tisk…

Anti-SLC32A1 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS7507
BWRLBS7507EA 10450 CZK
BWRLBS7507
Anti-SLC32A1 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human SLC32A1.

SLC32A1 pAb detects endogenous levels of SLC32A1 protein.

Type: Primary
Antigen: SLC32A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR