Tisk…

Anti-SMARCA5 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS7345
BWRLBS7345EA 10450 CZK
BWRLBS7345
Anti-SMARCA5 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human SMARCA5.

SMARCA5 pAb detects endogenous levels of SMARCA5 protein.

Type: Primary
Antigen: SMARCA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR