Tisk…

Anti-Rad50 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS7212
BWRLBS7212EA 10450 CZK
BWRLBS7212
Anti-Rad50 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human Rad50.

Rad50 pAb detects endogenous levels of Rad50 protein.

Type: Primary
Antigen: Rad50
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR