Tisk…

Anti-Catenin-alpha1/2 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS6976
BWRLBS6976EA 10450 CZK
BWRLBS6976
Anti-Catenin-alpha1/2 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human Catenin-alpha1.

Catenin-?1/2 pAb detects endogenous levels of Catenin-?1/2 protein.

Type: Primary
Antigen: Catenin-alpha1/2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR