Tisk…

Anti-STAR Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS6960
BWRLBS6960EA 10450 CZK
BWRLBS6960
Anti-STAR Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human STAR.

STAR pAb detects endogenous levels of STAR protein.

Type: Primary
Antigen: STAR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human; Mouse; Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR