Tisk…

Anti-Rpb1/POLR2A Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS6696
BWRLBS6696EA 10450 CZK
BWRLBS6696
Anti-Rpb1/POLR2A Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human Rpb1/POLR2A.

POLR2A pAb detects endogenous levels of POLR2A protein.

Type: Primary
Antigen: Rpb1/POLR2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR