Tisk…

Anti-MITF Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS6666
BWRLBS6666EA 10450 CZK
BWRLBS6666
Anti-MITF Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human MITF.

MITF pAb detects endogenous levels of MITF protein.

Type: Primary
Antigen: MITF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR