Tisk…

Anti-AMBRA1 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS6138
BWRLBS6138EA 10450 CZK
BWRLBS6138
Anti-AMBRA1 Rabbit Polyclonal Antibody
Protilátky
Recombinant full length Human AMBRA1.

AMBRA1 pAb detects endogenous levels of AMBRA1 protein.

Type: Primary
Antigen: AMBRA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR