Tisk…

Anti-RPA1 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS60446
BWRLBS60446EA 8030 CZK
BWRLBS60446
Anti-RPA1 Rabbit Polyclonal Antibody
Protilátky
A synthetic peptide corresponding to residues in Human RPA1.

RPA1 pAb detects endogenous levels of RPA1 protein.

Type: Primary
Antigen: RPA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR