Tisk…

Anti-Chemerin Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS60386
BWRLBS60386EA 8030 CZK
BWRLBS60386
Anti-Chemerin Rabbit Polyclonal Antibody
Protilátky
A synthetic peptide corresponding to residues in Human Chemerin.

Chemerin pAb detects endogenous levels of Chemerin protein.

Type: Primary
Antigen: Chemerin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR