Tisk…

Anti-IL28A Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS60330
BWRLBS60330EA 8030 CZK
BWRLBS60330
Anti-IL28A Rabbit Polyclonal Antibody
Protilátky
A synthetic peptide corresponding to residues in Human IL28A.

IL28A pAb detects endogenous levels of IL28A protein.

Type: Primary
Antigen: IL28A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR