Tisk…

Anti-ATP6AP1 Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS60265
BWRLBS60265EA 8030 CZK
BWRLBS60265
Anti-ATP6AP1 Rabbit Polyclonal Antibody
Protilátky
A synthetic peptide corresponding to residues in Human ATP6AP1.

ATP6AP1 pAb detects endogenous levels of ATP6AP1 protein.

Type: Primary
Antigen: ATP6AP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR