Tisk…

Anti-GK Rabbit Polyclonal Antibody

Dodavatel: Bioworld Technology
BS5733
BWRLBS5733EA 8030 CZK
BWRLBS5733
Anti-GK Rabbit Polyclonal Antibody
Protilátky
Synthetic peptide, corresponding to amino acids 500-545 of Human GK.

GK pAb detects endogenous levels of GK protein.

Type: Primary
Antigen: GK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR