Tisk…

Anti-Tau Rabbit Polyclonal Antibody [clone: S512]

Dodavatel: Bioworld Technology
BS3738
BWRLBS3738EA 8030 CZK
BWRLBS3738
Anti-Tau Rabbit Polyclonal Antibody [clone: S512]
Protilátky
Synthetic peptide, corresponding to amino acids 481-530 of Human Tau.

Tau (S512) pAb detects endogenous levels of Tau protein.

Type: Primary
Antigen: Tau
Clonality: Polyclonal
Clone: S512
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR