Tisk…

Anti-CK14 Rabbit Polyclonal Antibody [clone: Y151]

Dodavatel: Bioworld Technology
BS3500
BWRLBS3500EA 8030 CZK
BWRLBS3500
Anti-CK14 Rabbit Polyclonal Antibody [clone: Y151]
Protilátky
Synthetic peptide, corresponding to amino acids 111-160 of Human Cytokeratin 14.

Cytokeratin 14 (Y151) pAb detects endogenous levels of Cytokeratin 14 protein.

Type: Primary
Antigen: CK14
Clonality: Polyclonal
Clone: Y151
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR