Tisk…

Anti-Inhibin beta-C Rabbit Polyclonal Antibody [clone: V129]

Dodavatel: Bioworld Technology
BS3449
BWRLBS3449EA 8030 CZK
BWRLBS3449
Anti-Inhibin beta-C Rabbit Polyclonal Antibody [clone: V129]
Protilátky
Synthetic peptide, corresponding to amino acids 100-150 of Human Inhibin beta-C.

Inhibin ?-C (V129) pAb detects endogenous levels of Inhibin ?-C protein.

Type: Primary
Antigen: Inhibin beta-C
Clonality: Polyclonal
Clone: V129
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu Inhibin beta-C
Environmentally Preferable
Druh protilátky Primární
Klonovalita Polyclonal
Klon V129
Reaktivita Human
Hostitel Rabbit
Izotyp
Western blot Yes
Imunochemie Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR