Tisk…

Anti-CCHL Rabbit Polyclonal Antibody [clone: R104]

Dodavatel: Bioworld Technology
BS3019
BWRLBS3019EA 8030 CZK
BWRLBS3019
Anti-CCHL Rabbit Polyclonal Antibody [clone: R104]
Protilátky
Synthetic peptide, corresponding to amino acids 80-120 of Human CCHL.

CCHL (R104) pAb detects endogenous levels of CCHL protein.

Type: Primary
Antigen: CCHL
Clonality: Polyclonal
Clone: R104
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR