Tisk…

Anti-GPR174 Rabbit Polyclonal Antibody [clone: K167]

Dodavatel: Bioworld Technology
BS2779
BWRLBS2779EA 8030 CZK
BWRLBS2779
Anti-GPR174 Rabbit Polyclonal Antibody [clone: K167]
Protilátky
Synthetic peptide, corresponding to amino acids 132-178 of Human GPR174.

GPR174 (K167) pAb detects endogenous levels of GPR174 protein.

Type: Primary
Antigen: GPR174
Clonality: Polyclonal
Clone: K167
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR