Tisk…

Anti-Csk Rabbit Polyclonal Antibody [clone: N63]

Dodavatel: Bioworld Technology
BS2275
BWRLBS2275EA 8030 CZK
BWRLBS2275
Anti-Csk Rabbit Polyclonal Antibody [clone: N63]
Protilátky
Synthetic peptide, corresponding to amino acids N-terminus of Human Csk.

Csk (N63) pAb detects endogenous levels of Csk protein.

Type: Primary
Antigen: Csk
Clonality: Polyclonal
Clone: N63
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR