Tisk…

Anti-Hox-B9 Rabbit Polyclonal Antibody [clone: R237]

Dodavatel: Bioworld Technology
BS1940
BWRLBS1940EA 8030 CZK
BWRLBS1940
Anti-Hox-B9 Rabbit Polyclonal Antibody [clone: R237]
Protilátky
Synthetic peptide, corresponding to amino acids 200-250 of Human Hox-B9.

Hox-B9 (R237) pAb detects endogenous levels of Hox-B9 protein.

Type: Primary
Antigen: Hox-B9
Clonality: Polyclonal
Clone: R237
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR