Tisk…

Anti-HDAC6 Rabbit Polyclonal Antibody [clone: Q18]

Dodavatel: Bioworld Technology
BS1652
BWRLBS1652EA 8030 CZK
BWRLBS1652
Anti-HDAC6 Rabbit Polyclonal Antibody [clone: Q18]
Protilátky
Synthetic peptide, corresponding to the N-terminual of Human HDAC6.

HDAC6 (Q18) pAb detects endogenous levels of HDAC6 protein.

Type: Primary
Antigen: HDAC6
Clonality: Polyclonal
Clone: Q18
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR