Tisk…

Anti-PTEN Rabbit Polyclonal Antibody [clone: P374]

Dodavatel: Bioworld Technology
BS1304
BWRLBS1304EA 8030 CZK
BWRLBS1304
Anti-PTEN Rabbit Polyclonal Antibody [clone: P374]
Protilátky
Synthetic peptide, corresponding to amino acids 350-400 of Human PTEN.

PTEN (P374) pAb detects endogenous levels of PTEN protein.

Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone: P374
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR