Tisk…
Tubing couplings, rapid, NW 6,4 mm
Dodavatel: BURKLE
8761-1050 8771-1050 8760-2161 8772-1140 8761-1140 8771-1140 8761-1160 8771-1160 8761-1060 8771-1040 8771-1060 8761-1040 8760-2141 8771-2160 8761-1150 8771-1150 8760-1061 8761-2140 8771-2140 8763-1160 8775-1160 8763-1150 8775-2140 8760-1142 8761-2160 8763-1040 8773-1150 8775-1040 8770-1142 8763-1140 8770-1042 8772-1310 8773-1140 8763-2160 8770-2142 8770-1041 8773-1160 8762-1380 8772-1380 8760-1141 8762-1140 8760-1042
BURK8761-1050EA 512 CZK
BURK8761-1050 BURK8771-1050 BURK8760-2161 223-1133 BURK8761-1140 BURK8771-1140 BURK8761-1160 BURK8771-1160 BURK8761-1060 BURK8771-1040 BURK8771-1060 BURK8761-1040 BURK8760-2141 BURK8771-2160 BURK8761-1150 BURK8771-1150 BURK8760-1061 BURK8761-2140 BURK8771-2140 BURK8763-1160 BURK8775-1160 BURK8763-1150 BURK8775-2140 BURK8760-1142 BURK8761-2160 BURK8763-1040 BURK8773-1150 BURK8775-1040 BURK8770-1142 BURK8763-1140 BURK8770-1042 BURK8772-1310 BURK8773-1140 BURK8763-2160 BURK8770-2142 BURK8770-1041 BURK8773-1160 BURK8762-1380 BURK8772-1380 BURK8760-1141 BURK8762-1140 BURK8760-1042
Tubing couplings, rapid, NW 6,4 mm
Fitinky a konektory
Spring/lock Stainless steel V4A
Fast and safe connection for liquids, gases, compressed air and vacuum. Rapid couplings are the ideal when connecting and separating hoses, machines, apparatus, containers, etc. Top quality, with tested reliability, small space requirement, maximum possible flow, easy operation (single handed operation) and an elegant design distinguish the rapid couplings.The couplings are available in two different materials: POM (Polyoxymethylene) and PP (Polypropylene). The rapid couplings are available with a hose nozzle, hose screw joint, thread connector or as aMore Product Information