Tisk…
Buckets, Round, HDPE
Dodavatel: BURKLE
2306-0015 2306-0010
BURK2306-0015EA 1780 CZK
BURK2306-0015 BURK2306-0010
Buckets, Round, HDPE
Kbelíky
PE, white.
  • With contents scale, graduations 1000 ml
  • With spout
  • Handle made of metal
Laboratory bucket for transport, decanting and mixing. Safe emtying with wide spout.