Tisk…
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF647) [clone: BM-2]
Dodavatel: Biotium
BNC470062-500 BNC470062-100
BTIUBNC470062-500EA 15940 CZK
BTIUBNC470062-500 BTIUBNC470062-100
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF647) [clone: BM-2]
Protilátky
Recognizes 183 kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen. It reacts with myeloid precursor cells and granulocytes in bone marrow. Its antigen appears to be restricted to M2 and M3 acute myelogenous leukemia (AML) subtypes. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. This MAb reacts with early precursor and mature forms of human myeloid cells. It is useful in the identification of myelogenous leukemias, distinguishing granulocytic sarcomas from lymphoid malignancies and also inMore Product Information