Tisk…
Anti-Tyrosine Hydroxylase Rabbit Polyclonal Antibody
Dodavatel: Biosensis
R-148-50
BSENR-148-50EA 6980 CZK
BSENR-148-50
Anti-Tyrosine Hydroxylase Rabbit Polyclonal Antibody
Protilátky
Tyrosine hydroxylase (TH) is the rate-limiting enzyme in the synthesis of the catecholamines dopamine, epinephrine and norepinephrine. Therefore the regulation of the TH enzyme represents the central means for controlling the synthesis of these important catecholamines. FUNCTION: Plays an important role in the physiology of adrenergic neurons. CATALYTIC ACTIVITY: L-tyrosine + tetrahydrobiopterin + O2 = 3,4-dihydroxy-L-phenylalanine + 4a-hydroxytetrahydrobiopterin. COFACTOR: Fe(2+) ion. ENZYME REGULATION: Phosphorylation leads to an increase in the catalytic activity. PATHWAY: CatecholamineMore Product Information