Tisk…
Anti-beta NGF Chicken Polyclonal Antibody
Dodavatel: Biosensis
C-1558-500
BSENC-1558-500EA 9140 CZK
BSENC-1558-500
Anti-beta NGF Chicken Polyclonal Antibody
Protilátky
FUNCTION: Nerve growth factor is important for the development and maintenance of the sympathetic and sensory nervous systems. It stimulates division and differentiation of sympathetic and embryonic sensory neurons. SUBUNIT: Homodimer, associated by noncovalent forces. SUBCELLULAR LOCATION: Secreted protein. SIMILARITY: Belongs to the NGF-beta family.
Antibody recognizes NGF in human, mouse and rat and likely other species as well as NGF is highly conserved but they have not been tested. Approved applications include: ELISA, WB and IHC (frozen, non-paraffin embedded tissues tested); notMore Product Information