Tisk…
Anti-Orexin B Chicken Polyclonal Antibody
Dodavatel: Biosensis
C-1532-100
BSENC-1532-100EA 9140 CZK
BSENC-1532-100
Anti-Orexin B Chicken Polyclonal Antibody
Protilátky
FUNCTION: Neuropeptides that play a significant role in the regulation of food intake and sleep-wakefulness, possibly by coordinating the complex behavioral and physiologic responses of these complementary homeostatic functions. A broader role in the homeostatic regulation of energy metabolism, autonomic function, hormonal balance and the regulation of body fluids, is also suggested. Orexin-A binds to both OX1R and OX2R with a high affinity, whereas orexin-B binds only to OX2R with a similar high affinity. SUBCELLULAR LOCATION: Endoplasmic reticulum; rough endoplasmic reticulum. AssociatedMore Product Information