Tisk…

Anti-MBD2 Polyclonal Antibody

Dodavatel: Boster Bio
RP1049
BSBTRP1049EA 15720 CZK
BSBTRP1049
Anti-MBD2 Polyclonal Antibody
Protilátky
Polyclonal antibody for MBD2 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: WB. Reactive species: Human. MBD2 information: Molecular Weight: 43255 MW; Subcellular Localization: Nucleus . Nuclear, in discrete foci. Detected at replication foci in late S phase; Tissue Specificity: Highly expressed in brain, heart, kidney, stomach, testis and placenta.

Type: Primary
Antigen: MBD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR