Tisk…
HCCA matrix
Dodavatel: BRUKER DALTRONIK
8255344
BRUK8255344EA 0 CZK
BRUK8255344
HCCA matrix
Systémy pro purifikaci proteinů Proteinové čistící reagence
Pre-portioned HCCA matrix provides easy and convenient preparation of HCCA matrix solutions.
  • Stable and easy to handle for sample preparation
HCCA has been developed especially for Bruker MALDI instruments and purified to ensure accurate results and maintain system cleanliness. This reduces frequency of preventive maintenance cleans required.