Tisk…
BROEN-LAB UniFlex™ Flexible Stainless Steel Pipes for Special Water
Dodavatel: BROEN-LAB
93Z40401000
BROE93Z40401000EA 0 CZK
BROE93Z40401000
BROEN-LAB UniFlex™ Flexible Stainless Steel Pipes for Special Water
Fitinky a konektory
BROEN-LAB offers BROEN-LAB UniFlex™ hoses and flexible Stainless Steel pipes as an easy way to make the installation in the laboratory where safety and efficiency are required. BROEN-LAB UniFlex™ hoses are the ideal solution where there is a higher requirement for flexibility. Installation with flexible hoses saves time and labour - in other words - reduced installation costs.
  • Inner hose material: Stainless steel 1,4404 similar to AISI 316 L
  • Braiding: None
  • Materials with media contact: Stainless steel 1.4404, EPDM, aramide fibre/NBR gasket