Tisk…
Bottle-top dispensers, Dispensette® S Organic, DAkkS
Dodavatel: Brand
DAKKS4630161 DAKKS4630361 DAKKS4630360 DAKKS4630151 DAKKS4630341 DAKKS4630351
BRNDDAKKS4630161EA 20160 CZK
BRNDDAKKS4630161 BRNDDAKKS4630361 BRNDDAKKS4630360 BRNDDAKKS4630151 BRNDDAKKS4630341 BRNDDAKKS4630351
Bottle-top dispensers, Dispensette® S Organic, DAkkS
Dávkovače na lahve
Analogue-adjustable or Digital Easy Calibration.
  • With or without recirculation valve
  • Autoclavable at 121 °C
  • Model of volume adjustment available: Analog (Analog volume display, fast volume adjustment; simple calibration adjustment with supplied tool)
  • Digital EASY CALIBRATION: Easy to read digital display; dispensing volume can be set accurately and reproducibly. Easy Calibration technique for adjustment in seconds without tools; alteration of factory setting is automatically indicated
  • DE-M marking
  • Easy to calibrate and adjust in order to comply with ISO 900
More Product Information