Tisk…
Shaking and Heating Module Teleshake 95 for Liquid Handling Station
Dodavatel: Brand
709470
BRND709470EA 103870 CZK
BRND709470
Shaking and Heating Module Teleshake 95 for Liquid Handling Station
Automated Liquid Handling Systems
With the Teleshake 95 module, shaking and heating of samples in microplates and PCR consumables is possible directly inside the Liquid Handling Station pipetting robot from BRAND, without removing the plates from the robot.
  • For shaking and heating 96-well and 384-well microplates and PCR plates
  • Shaking frequency range 100 - 1900 rpm
  • Max temperature 90 °C
Saves time and reduces the risk of sample contamination. Applications include sample incubation and intensive stirring of samples during reaction at room temperature or higher. Thanks to its modular
More Product Information