Tisk…
Magicard™ Colour Film for Magicard™ 300, 300 Shot
Dodavatel: Brady
321973
BRDY321973EA 2640 CZK
BRDY321973
Magicard™ Colour Film for Magicard™ 300, 300 Shot
Pásky do tiskáren
The Magicard™ colour ribbon is perfect for full colour prints with a Magicard™ 300 printer.
  • High-quality, full colour ribbon
  • Yellow, Magenta, Cyan, Black with an overcoat
  • Includes a cleaning roller for dust free printing
Print 300 single-sided cards.
Informace o dodávce: Includes 1 dye film and 1 cleaning roller.