Tisk…
Destructible Vinyl Tamper Resistant Labels for M710, M610 and M611
Dodavatel: Brady
312075 312074 173867 312101
BRDY312075EA 1700 CZK
BRDY312075 BRDY312074 BRDY173867 BRDY312101
Destructible Vinyl Tamper Resistant Labels for M710, M610 and M611
Štítky
Labels for the M610 and M611 Thermal Transfer Portable Printers. These parts can also be printed on the M710 Label Printer provided the roll adapter (M71-ADAPT) is used.
  • High performance white matte vinyl with permanent adhesive
  • Ideal for applications that require resistance to product tampering
  • Use caution when removing from liner as material is fragile
  • Service temperature of −40 to 82 °C (−40 to 180 °F)
Ideal for security identification, general identification, and laboratory labelling applications.
A ribbon is required to print this material.More Product Information