Tisk…
Engraved Replacement Labels for M610, M611
Dodavatel: Brady
312041 312038 312033 312037 312039 312036 312034 312032 312031 312040 312035
BRDY312041EA 13740 CZK
BRDY312041 BRDY312038 BRDY312033 BRDY312037 BRDY312039 BRDY312036 BRDY312034 BRDY312032 BRDY312031 BRDY312040 BRDY312035
Engraved Replacement Labels for M610, M611
Štítky
Engraved plate replacement labels for the M610 Portable Printer.
  • Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags
  • Polyethylene foam laminate Polyester material
  • Service temperature of −40 to +100 °C
  • Gloss finish
Ideal for data and telecommunications labelling, electrical labelling, and general identification applications.
Balení: 300 labels per box.